Learn More
CPC Scientific H-Arg-Ser-Gly-Pro-Pro-Gly-Leu-Gln-Gly-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Ala-Ser-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Met-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OREX002B
Encompass
Hypothalamic neuropeptide that regulates feeding behavior and significantly augments food intake in a dose-dependent manner when a bolus is administered intracerebroventricularly. After a 48-hour fast, hypothalamic prpro-orexin mRNA level is up-regulated 2.4. The discovery of orexins and their receptors may provide a novel molecular basis for understanding the role of the lateral hypothalamic areas in the regulation of energy homeostasis.SEQUENCE: H-Arg-Ser-Gly-Pro-Pro-Gly-Leu-Gln-Gly-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Ala-Ser-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Met-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: RSGPPGLQGRLQRLLQASGNHAAGILTM-NH2MOLECULAR FORMULA: C123H212N44O35S1MOLECULAR WEIGHT: 2898.39STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [205640-91-1]SYNONYMS: Hypocretin-2 (human)RESEARCH AREA: ObesityREFERENCES:M.W. Schwartz, Nat. Med., 4, 385 (1998)T. Sakurai et al., Cell, 92, 573 (1998) SKU(s): OREX-002A, OREX-002B
Catalog No. 50-210-9996
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.