Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Orexin B Recombinant Rabbit Monoclonal Antibody (2O8I1)

Catalog No. MA558091
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA558091 20 μL
MA558092 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA558091 Supplier Invitrogen™ Supplier No. MA558091
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MNLPSTKVSW AAVTLLLLLL LLPPALLSSG AAAQPLPDCC RQKTCSCRLY ELLHGAGNHA AGILTLGKRR SGPPGLQGRL QRLLQASGNH AAGILTMGRR.

Orexin B is a neuropeptides that play a significant role in the regulation of food intake and sleep-wakefulness, possibly by coordinating the complex behavioral and physiologic responses of these complementary homeostatic functions. A broader role in the homeostatic regulation of energy metabolism, autonomic function, hormonal balance and the regulation of body fluids, is also suggested. Orexin-A binds to both OX1R and OX2R with a high affinity, whereas orexin-B binds only to OX2R with a similar high affinity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Orexin B
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 2O8I1
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Hcrt
Gene Accession No. O43612
Gene Alias HCRT; Hcrt1; Hcrt2; Hypocretin; hypocretin (orexin) neuropeptide; hypocretin neuropeptide precursor; Hypocretin-1; Hypocretin-2; NRCLP1; orexin; orexin A; orexin B; orexin-A; Orexin-B; ORX; Ox; PPORX; Ppox; prepro-orexin
Gene Symbols Hcrt
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3060
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.