Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Org 27569, Tocris Bioscience™
Description
Potent CB1 receptor allosteric modulator (pEC50 = 8.24). Significantly increases binding of the CB1 agonist [3H]CP 55.940 (pKb = 5.67) and decreases binding of the CB1 inverse agonist [3H]SR 141716A (pKb = 5.95). Inhibits CB1 receptor antagonist efficacy in vitro (pKb = 7.57).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 409.95 |
| Percent Purity | >99% |
| Chemical Name or Material | Org 27569 |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?