Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Organic Carbon Standard, 10,000 ppm C (Ethylene Glycol in 0.2% H2SO4), Ricca Chemical
SDP

Catalog No. p-4770347 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R1849950500 500 mL Amber Glass Bottle
R18499504C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R1849950500 Supplier Ricca Chemical Company Supplier No. 1849.9516
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 107-21-1 , 7664-93-9
Molecular Formula C8H5KO4
Molecular Weight (g/mol) Mixture
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Packaging Amber Glass Bottle
Odor Odorless
Assay Percent Range 10, 000 ± 50ppm
Physical Form Liquid
Color Colorless to Yellow
Melting Point Approximately 0°C
Solubility Infinitely soluble in water
Type Standard
Solvent or Matrix Water
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Organic Carbon Standard, 10,000mg/L C, 10,000ppm C (Ethylene Glycol in 0.2% Sulfuric Acid)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.