Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Organic Vapor, Acid Gas Cartridge with HEPA Filter

Catalog No. 19185706
Change view
Click to view available options
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-185-706 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-185-706 Supplier Honeywell™ Supplier No. PA7OVAGHE

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy

The Organic Vapor, Acid Gas Cartridge with HEPA filter is designed to be used with the PA700 PAPR.

  • Organic Vapor, Acid Gas [Chlorine, Hydrogen Chloride, Sulfur Dioxide, Hydrogen Fluoride, Chlorine Dioxide, Hydrogen Sulfide], with High Efficiency Particulate Air-Purifying (HEPA) filter
TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type OV-AG
Color Yellow
Product Type PAPR Filter Cartridge
For Use With (Equipment) Honeywell PA700 Powered Air Purifying Respirator (PAPR)
Model PA700 PAPR Series Filter Cartridge
Quantity 4/Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.