Learn More
Dentec Safety™ Organic Vapor Cartridge with N95 Prefilters and Retainers
Shop All Dentec Safety Specialists Inc. Products
Description
- Unique space plenums allow users to access the entire filter material reducing breathing resistance, providing longer cartrdige life, reducing costs and improving productivity.
- Offset connector allows the user to rotate the cartridge into 3 different positions improving visibility
- Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
Specifications
Specifications
| Material | Plastic |
| Product Type | Respirator Accessories |
| For Use With (Application) | Lacquer and enamel paints, Pesticides, Welding, Dusts and Non-oil mists |
| For Use With (Equipment) | Masks |
| Includes | Cartridges and filters |
| Quantity | 12 Pairs |