Learn More
Organochlorine Pesticide Mix C #1, Restek™
SDP
Catalog No. 06719912 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
06-719-912 1 mL
1 options
Catalog No. 06-719-912 Supplier Restek Corporation Supplier No. 32296
Compounds:
  • Chlorobenzilate (510-15-6)
  • Diallate (cis & trans) (2303-16-4)
  • 1,2-Dibromo-3-chloropropane (96-12-8)
  • Hexachlorobenzene (118-74-1)
  • Hexachlorocyclopentadiene (77-47-4)
  • Isodrin (465-73-6)
Specifications
Packaging Ampul
Concentration 200 μg/mL each in hexane:toluene (1:1)
Type Multi-component Standard
Quantity 1 mL
ShelfLife 6 months