Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Organochlorine Pesticide Mixture, 1000μg/mL in hexane/toluene, Ultra Scientific

Catalog No. USPPM808C1
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USPPM-808C-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USPPM-808C-1 Supplier Agilent Technologies Supplier No. PPM808C1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Organochlorine Pesticide Mixture
CAS Max % 49.67, 0.03, 0.03, 0.03, 0.03
CAS 108-88-3, 309-00-2, 319-84-6, 319-85-7, 319-86-8
Analyte or Component Names n-hexane,Toluene,Aldrin
Element n-hexane, Toluene, Aldrin
Solvent or Matrix Hexane/Toluene
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.