Learn More
FUJIFILM IRVINE SCIENTIFIC INC IS Giant Cell Tumor-Conditioned Medium, 50 mL
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 9100650ML
Encompass
IS Giant Cell Tumor-Conditioned Medium is a unique conditioned medium derived from a human tumor cell line, IS GCT-CM provides a winning combination of GM-CSF, G-CSF, IL-1 and IL-6, to ensure dependable performance in a wide range of applications. This liquid medium easily grows hematopoietic cells, improves bone marrow cytogenetic analysis, isolates and recovers HIV, and produces and grows human hybridomas.
Catalog No. NC9819231
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.