Learn More
Thermo Scientific™ Orion™ 8103BNUWP ROSS Ultra™ pH Electrode PROMO
Description
- Superior measurement stability, faster response, greater accuracy, longer life, least long-term drift and more reproducible results
- Featured with WP- waterproof BNC connectors
Includes:
- 60 mL bottle of Orion ROSS electrode fill solution 810007
Requires:
- pH buffers for calibration
- Beakers, plastic or glass
- Magnetic stirrer or Orion stirrer probe
- Distilled or deionized water
Compatible with:
Waterproof BNC connector is compatible with any standard BNC connector both on Thermo Scientific™ Orion Star™ Series meters and competitive meter models.
Recommended for:
- Clinical, pharmaceutical, and food labs, wherever sample size is a constraint
Specifications
Specifications
| Description | Semi-micro tipped glass combination electrode; 165 x 6 mm |
| Connector Types | BNC |
| Junction | Ceramic |
| Material | Glass Body |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 100°C |
| Type | pH Electrode |
| Filling Solution Type | 3M KCl |
| Includes | 60 mL bottle of Orion ROSS electrode fill solution |
| Internal Reference | ROSS |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.