Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ AQ3700 AQUAfast™ Colorimeter

Catalog No. 13642400
Change view
Click to view available options
Type:
Colorimeter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-642-400 Colorimeter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-642-400 Supplier Thermo Scientific™ Supplier No. AQ3700
Add to Cart
Add to Cart

Select this multiparameter colorimeter to manage your water and wastewater analysis using over 90 preprogrammed methods.

Select the Thermo Scientific™ Orion™ AQUAfast AQ3700 Colorimeter to manage your water and wastewater analysis needs. This instrument has over 90 pre-programmed methods for common test parameters with automatic selection of 6 LED wavelengths and countdown timer. Methods include total nitrogen and total phosphate nutrient testing or create up to 10 custom methods using 2-14 standards and blank. This IP67 waterproof meter records 1,000 data points, offers a long battery life of about 26 hours continuous use or 3,500 tests, and multi-language user interface in English, French, German, Italian, Polish, Portuguese and Spanish.

Features:

  • Wide range of disinfectants in various formats to broaden our offering to a variety of industries
  • Large graphic display with step-by-step user instructions
  • Automatic countdown to measurement
  • Consistent results from shift to shift
  • Selectable languages of English, French, German, Italian, Spanish, Portuguese (Brazilian), and Polish with other languages uploadable via RJ 45 connection

Requires:

  • 4 AA batteries

Recommended for:

  • Wastewater
  • Drinking water
  • Nutrient monitoring
  • Environmental testing
  • Food and beverage
  • Industrial applications
Note: For the most accurate results, use only Thermo Scientific Orion reagents.
TRUSTED_SUSTAINABILITY

Specifications

Accuracy 2% Full scale
Backlight Option Yes
Certifications/Compliance FCC Part 15; CE
Cuvette Details Round 13 mm, 16 mm, 24 mm & 16 mm COD Digestion Tubes
Battery Type AA
Display Graphic
Languages English, French, German, Spanish, Italian
Light Source 6 x LED
Product Line AQUAfast
Type Colorimeter
Wavelength Range 430, 530, 560, 580, 610, 660 nm
Includes AQ3700 colorimeter, 4 batteries (Type AA/LR 6); Round 24 mm vials with caps; Round 16 mm vials with caps; 16 mm vial adapter; 13 mm vial adapter; Vial cleaning brush; Stirring rod
Interface Internal RJ45 port
Keypad Numeric keys, ON/OFF, Up, Down, Enter, Timer, Zero, Test, Cal, Mode, Store, Shift, plus 3 Function keys
Outputs IRDA
Reagent Form Powder, tablet, liquid, reagent tubes, digestion tubes. Color, Turbidity and Suspended Solids tests do not require reagents
Resolution ±0.005 A
Sample Size 10 mL
Storage Requirements 5°C to 40°C
Test Method Up to 90 and 10 user defined programmed methods
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.