Learn More
Thermo Scientific™ Orion™ AQUAfast™ Free & Total Chlorine Bulk DPD Powder Reagents
Catalog No. 13301154
Change view
Click to view available options
Includes:
AQ250F Dispenser, 1 vial
AQ250T Dispenser, 1 vial
DPD Powder Refill for AQ250F
DPD Powder Refill for AQ250T
Quantity:
2 x 250 count
250 count
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Quantity
13-301-154 DPD Powder Refill for AQ250F 250 count
13-301-153 AQ250F Dispenser, 1 vial 250 count
13-301-155 DPD Powder Refill for AQ250F 2 x 250 count
13-301-156 AQ250T Dispenser, 1 vial 250 count
13-301-157 DPD Powder Refill for AQ250T 250 count
13-301-158 DPD Powder Refill for AQ250T 2 x 250 count
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 13-301-154 Supplier Thermo Scientific™ Supplier No. AC71P1
Add to Cart
Add to Cart
Thermo Scientific™ Orion™ AQUAfast Free and Total Chlorine Bulk DPD Powder Dispensers and Vials are designed for high volume testing of residual chlorine in water samples.
Measure residual chlorine in water samples using Thermo Scientific™ Orion™ AQUAfast Free and Total Chlorine Bulk DPD Powder Dispensers and Vials with Orion AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. The powder chemistries dissolve quickly in the solution and are easy to mix. Each vial contains 250 counts of DPD reagent and is available for free chlorine or total chlorine analysis in the lab or field.
  • Measure 0.02 to 2.00 mg/L (ppm) free chlorine or total chlorine
  • Bulk powder dispensers are designed for high sample throughput – reduces the cost per test and minimizes waste compared with individually packaged reagents 
  • Equivalent to other widely used and recognized chemistries including Orion AQUAfast AC4P71 free chlorine and Orion AQUAfast AC4P72 total chlorine powder packs
  • Compatible with Orion AQUAfast AQ4000, AQ3700 and AQ3170 colorimeters
  • Compatible with Orion AquaMate AQ8000 and AQ7000 spectrophotometers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) AQUAfast AQ250F free chlorine DPD bulk powder dispenser
Includes DPD Powder Refill for AQ250F
Quantity 250 count
Range 0.02 to 2 mg/L Cl2
Reagent Form Powder
Reagents Chlorine
Test Method DPD