Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits

Catalog No. 13642018
Change view
Click to view available options
Type:
Alkalinity
Aluminum
Ammonia
Chloride
Chlorine
Copper
Iron
Nitrate
Nitrite
Phosphate
Silica
Sulfate
Sulfide
Zinc
pH
Range:
0.01 to 0.3 mg/L Al
0.01 to 0.5 mg/L N
0.01 to 6 mg/L Cl2
0.02 to 1 mg/L Fe
0.02 to 1 mg/L N
0.02 to 1 mg/L Zn
0.04 to 0.5 mg/L S
0.05 to 4 mg/L PO4
0.05 to 4 mg/L SiO2
0.05 to 5 mg/L Cu
0.5 to 25 mg/L Cl-
1 to 40 mg/L
1 to 80 mg/L PO4
5 to 100 mg/L SO4
5 to 200 mg/L CaCO3
6.5 to 8.4 pH
Quantity:
100 DPD No 1 Tablets
100 DPD No 4 Tablets
100 Tablets
100 Tests
50 Tests
50 Tests Each (free and total)
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Range Quantity
13-642-018 Phosphate 1 to 80 mg/L PO4 50 Tests
13-642-000 Alkalinity 5 to 200 mg/L CaCO3 100 Tests
13-642-001 Aluminum 0.01 to 0.3 mg/L Al 50 Tests
13-642-012 Ammonia 0.02 to 1 mg/L N 50 Tests
13-642-003 Chloride 0.5 to 25 mg/L Cl- 50 Tests
13-642-004 Chlorine 0.01 to 6 mg/L Cl2 100 DPD No 1 Tablets
13-642-689 Chlorine 0.01 to 6 mg/L Cl2 50 Tests Each (free and total)
13-642-005 Chlorine 0.01 to 6 mg/L Cl2 100 DPD No 4 Tablets
13-642-007 Copper 0.05 to 5 mg/L Cu 50 Tests
13-642-023 Zinc 0.02 to 1 mg/L Zn 50 Tests
13-642-010 Iron 0.02 to 1 mg/L Fe 100 Tests
13-642-014 Nitrite 0.01 to 0.5 mg/L N 100 Tests
13-644-937 pH 6.5 to 8.4 pH 100 Tests
13-642-017 Phosphate 0.05 to 4 mg/L PO4 50 Tests
13-642-019 Silica 0.05 to 4 mg/L SiO2 50 Tests
13-644-938 Silica 0.05 to 4 mg/L SiO2 100 Tablets
13-642-021 Sulfate 5 to 100 mg/L SO4 100 Tests
13-642-022 Sulfide 0.04 to 0.5 mg/L S 100 Tests
13-642-013 Nitrate 1 to 40 mg/L 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Catalog No. 13-642-018 Supplier Thermo Scientific™ Supplier No. AC2096
Add to Cart
Add to Cart

Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.

Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.

  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
  • No. of tests: 50 to 100
TRUSTED_SUSTAINABILITY

Specifications

Type Phosphate
Test Reagent Formats Tablet
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast
Description AQUAfast phosphate (ortho) HR reagent tablets, 50 tests
Includes Set of 2: 50 Phosphate No 1 HR Tablets, 50 Phosphate No 2 HR Tablets
Method Vanado-Molybdate
For Use With (Application) Colorimetric water and wastewater testing equipment
Recommended Accessories AQUAfast 24mm round vials (Cat. No. AC2V24)
Range 1 to 80 mg/L PO4
Quantity 50 Tests

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.