Learn More
Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits
Description
Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.
- For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
- No. of tests: 50 to 100
Specifications
Specifications
| Type | Sulfide |
| Test Reagent Formats | Tablet |
| For Use With (Equipment) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast |
| Description | AQUAfast sulfide reagent tablets, 100 tests |
| Includes | Set of 2: 100 Sulfide No 1 Tablets, 100 Sulfide No 2 |
| Method | DPD/Catalyst |
| For Use With (Application) | Colorimetric water and wastewater testing equipment |
| Recommended Accessories | AQUAfast 24mm round vials (Cat. No. AC2V24) |
| Range | 0.04 to 0.5 mg/L S |
| Quantity | 100 Tests |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.