Learn More
Thermo Scientific™ Orion™ Ionplus™ Sure-Flow™ Bromide Combination Ion Selective Electrode
Catalog No. 1329989
Change view
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13-299-89 BNC
13-645-158 BNC (Waterproof)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-299-89 Supplier Thermo Scientific™ Supplier No. 9435BN
Measure bromide ions in aqueous solutions quickly, simply, accurately and economically.
Measure bromide ions in aqueous solutions quickly, simply and economically with the Thermo Scientific™ Orion™ Bromide Electrodes.
Bromide half-cell electrodes, Part Numbers  9435BN and 9435SC, when used in conjunction with the Thermo Scientific™ Orion™ 900200 double junction reference electrode offers the benefits of the Sure-Flow junction design.
Bromide combination electrode, Part Number 9635BNWP, does not require a separate reference electrode making it convenient to use with small sample volumes.
Related Products
  • Calibration standards: 943506 (0.1 M NaBr)
  • Required ISA: 940011
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC
Description With BNC connector
Junction Sure-Flow Junction
For Use With (Equipment) ISE Meter
Type Solid state half-cell