Learn More
Thermo Scientific™ Orion™ Electrode Adapter Cables
Catalog No. 0935771
Click to view available options
Connector Types:
BNC
Screw Cap
Meter Input:
2 mm Pin Tip
BNC
E DIN (Knick, Schott, WTW)
Phillips
Radiometer No. 7
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
  • For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
  • Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC
Description E DIN (Knick, Schott, WTW); BNC; BNC to E DIN Adapter
For Use With (Equipment) Meters That Have BNC
Type Electrode Cable
Meter Input E DIN (Knick, Schott, WTW)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.