Learn More
Thermo Scientific™ Orion™ Electrode Adapter Cables
Description
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
- For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
- Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
Specifications
Specifications
| Connector Types | BNC |
| Description | Radiometer No. 7; BNC; BNC to Radiometer Adapter |
| For Use With (Equipment) | Meters with 2 mm Reference Pins Connectors, Meters with BNC Connectors, Meters with British Connectors, Meters with Certain LEMO Connectors, Meters with E DIN Connectors, Meters with Radiometer Connectors, Meters with Stripped Lead Inputs Connectors, Meters with US Standard Connectors |
| Type | Electrode Adapter Cable |
| Meter Input | Radiometer No. 7 |