Learn More
Thermo Scientific™ Orion™ Gas-Sensing ISE membranes
Description
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Fresh membranes provide optimum performance with gas-sensing ion selective electrodes. Ammonia electrodes require different membranes if using standard electrode or high performance ammonia electrode.
Specifications
Specifications
| Description | Gas-Sensing ISE membrane |
| For Use With (Equipment) | Standard Ammonia Electrode, 9512BNWP, 9512 |
| Type | Gas-Sensing ISE Membrane |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.