Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents

Catalog No. 13641852
Change view
Click to view available options
Type:
Acid Reagent
Alkaline Reagent
CISA
ISA Solution
Iodide Reagent
Ionic Strength Adjustor Buffer
Nitrate Interference Suppressor
Reconditioning Solution
Storage Solution
Sulfide Anti-Oxidant Buffer
TISAB II
TISAB III
pH Adjusting ISA
pH Adjusting Low Level ISA
For Use With (Equipment):
Ammonia Electrode
Bromide Electrode, Cadmium Electrode, Chloride Electrode, Cupric Electrode, Iodide Electrode, Sufide Electrode, Thiocyanate Electrode
Calcium Electrode
Carbon Dioxide Electrode
Chloride Electrode
Chlorine Electrode
Cyanide Electrode
Fluoride Electrode
Fluoroborate Electrode, Nitrate Electrode, Perchlorate Electrode
Nitrate Electrode
Potassium Electrode
Sodium Electrode
Sodium ISE
Sulfide Electrode
Quantity:
475 mL
5 x 50 mL
1 Gal.
4 x 475 mL
2 x 1 L
4 x 1 Gal.
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment) Quantity
13-641-852 ISA Solution Bromide Electrode, Cadmium Electrode, Chloride Electrode, Cupric Electrode, Iodide Electrode, Sufide Electrode, Thiocyanate Electrode 475 mL
13-641-743 Storage Solution Sodium Electrode 475 mL
13-641-748 Ionic Strength Adjustor Buffer Sodium ISE 475 mL
13-641-749 Reconditioning Solution Sodium Electrode 475 mL
13-641-802 Nitrate Interference Suppressor Nitrate Electrode 475 mL
13-641-850 ISA Solution Fluoroborate Electrode, Nitrate Electrode, Perchlorate Electrode 475 mL
13-641-927 ISA Solution Potassium Electrode 475 mL
13-641-851 ISA Solution Calcium Electrode 475 mL
13-642-578 TISAB II Fluoride Electrode 1 Gal.
13-641-874 TISAB III Fluoride Electrode 475 mL
13-641-873 TISAB II Fluoride Electrode 4 x 1 Gal.
13-641-882 Sulfide Anti-Oxidant Buffer Sulfide Electrode 4 x 475 mL
13-641-878 CISA Chloride Electrode 2 x 1 L
13-641-770 ISA Solution Carbon Dioxide Electrode 475 mL
13-641-853 Alkaline Reagent Cyanide Electrode 475 mL
15-200-960 pH Adjusting Low Level ISA Ammonia Electrode 475 mL
13-641-883 pH Adjusting ISA Ammonia Electrode 475 mL
13-641-823 Iodide Reagent Chlorine Electrode 5 x 50 mL
13-641-854 Acid Reagent Chlorine Electrode 475 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Catalog No. 13-641-852 Supplier Thermo Scientific™ Supplier No. 940011
Add to Cart
Add to Cart

Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents.

Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents. Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.

Warranty: Out of box

Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements. The ammonia pH-adjusting ISA (951211) and low-level ammonia pH-adjusting ISA (951210) both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required. The ammonia pH-adjusting ISA (951211) prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results. The low-level ammonia pH-adjusting ISA (951210) is ideal for ammonia samples with a concentration of 10ppm and below that do not contain metal interferences.

All ionic strength adjusters and special reagents are packaged as one 1 pint (475mL) bottle, except:

  • Cat. No. 941709 is packaged as two 1 quart (2 x 1L) bottles
  • Cat. No. 941609 is packaged as four 1 pint (4 x 475mL) bottles
  • Cat. No. 940909 is packaged as one 1 gallon (3.7L) bottle
  • Cat. No. 940999 is packaged as four 1 gallon (4 x 3.7L) bottles
TRUSTED_SUSTAINABILITY

Specifications

Description ISA for Bromide, Cadmium, Chloride, Cupric, Iodide, Sufide, Thiocyanate Electrodes
For Use With (Equipment) Bromide Electrode, Cadmium Electrode, Chloride Electrode, Cupric Electrode, Iodide Electrode, Sufide Electrode, Thiocyanate Electrode
Type ISA Solution
For Use With (Application) Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes
Packaging Bottle
Quantity 475 mL

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.