Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ PerpHecT™ ROSS™ Combination pH Micro Electrode PROMO

Catalog No. 13642800PMO
Change view
Click to view available options
Connector Types:
BNC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13-642-800PMO BNC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-642-800PMO Supplier Thermo Scientific™ Supplier No. 8220BNWPKIT

When you purchase a Thermo Scientific™ ROSS™ pH Electrode, you’ll receive a ROSS Buffer Kit at no additional cost. Order using the catalog numbers below and your products will ship together.

Includes:

  • 1 m cable

Requires:

  • pH Meter
  • Electrode
  • Beakers - plastic or glass
  • Magnetic stirrer
  • Required solutions
  • Two pH buffers: pH 7 and pH 4 /10
  • ROSS internal filling solution: 3M KCl; Orion Cat. No. 810007

Compatible with:

  • Orion™ PerpHecT™ meters
TRUSTED_SUSTAINABILITY

Specifications

Description Combination pH Micro Electrode
Connector Types BNC
Junction Double
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Drift Less than 0.002 pH units/day
Electrode Glass double junction; Refillable
Internal Reference ROSS
Min. Immersion 4.5 mm
Storage Requirements To ensure a quick response and free-flowing liquid junction the sensing element and reference junction must not be allowed to dry out. Short-term Storage (up to one week): Soak electrode in ROSS pH Electrode Storage Solution, Orion 810001 or use pH Electrode Storage Solution, Orion 910001. To minimize the chance of breakage the 8220BNWP should be attached to an electrode stand and suspended in a beaker that contains pH electrode storage solution. The electrode should not touch the sides or bottom of the beaker. Long-term Storage (more than one week): The reference chamber should be filled and the filling hole securely covered. Cover the sensing element and reference Junction with the protective cap/sleeve containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode.
Length (Metric) 155 mm
Length (Metric) Cable 1 m
Diameter (Metric) Cap 3 mm
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.