Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ pH Buffer Bottles

Catalog No. 0935769
Change view
Click to view available options
Quantity:
475 mL
5 x 60 mL
pH:
1.68 ± 0.01 pH
10.01 ± 0.02 pH
12.46 ± 0.03 pH
4.01 ± 0.01 pH
5.00 ± 0.02 pH
6.86 ± 0.01 pH
7.00 ± 0.01 pH
9.18 ± 0.01 pH
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity pH
09-357-69 5 x 60 mL 4.01 ± 0.01 pH
13-643-002 475 mL 1.68 ± 0.01 pH
13-643-007 5 x 60 mL 1.68 ± 0.01 pH
13-079-319 475 mL 10.01 ± 0.02 pH
09-357-68 5 x 60 mL 10.01 ± 0.02 pH
13-643-001 475 mL 12.46 ± 0.03 pH
13-643-005 5 x 60 mL 12.46 ± 0.03 pH
13-301-133 475 mL 4.01 ± 0.01 pH
13-640-8 475 mL 5.00 ± 0.02 pH
13-643-003 475 mL 6.86 ± 0.01 pH
13-643-006 5 x 60 mL 6.86 ± 0.01 pH
13-641-857 475 mL 7.00 ± 0.01 pH
09-357-70 5 x 60 mL 7.00 ± 0.01 pH
13-643-004 475 mL 9.18 ± 0.01 pH
13-643-008 5 x 60 mL 9.18 ± 0.01 pH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 09-357-69 Supplier Thermo Scientific™ Supplier No. 910460
Add to Cart
Add to Cart
This item is not returnable. View return policy

Perform accurate, reproducible pH calibrations with these color-coded buffer bottles.

Use Thermo Scientific™ Orion™ pH buffer bottles to perform accurate, reproducible pH calibrations on your pH meter and pH electrode measurement system. Orion pH buffers are available in 475mL bottles for everyday use and laboratory and bench pH analysis and available in 60 mL bottle 5 packs for occasional use and portable and field pH analysis. Using high-quality pH buffers to calibrate your pH system is the best way to ensure your pH measurements are correct and repeatable.

All Orion pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations and NIST traceability. Manufactured under ISO 9000 quality standards.

Buffer Color Codes:

  • pH 4.01 buffer is pink
  • pH 5.00 buffer is orange
  • pH 7.00 buffer is yellow
  • pH 10.01 buffer is blue

The buffers are available in pint (475mL) bottles, portable five packs of 2 oz. (5 x 60mL) bottles, convenient All-in-One buffer kits, 5 gallon (19L) Cubitainers™ and single-use pouches.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material pH Buffer, bottles
Color Pink
Description Orion pH 4.01 buffer, color-coded pink, 5x60mL bottles
For Use With (Application) Accurate pH calibrations and NIST traceability, portable and field analysis
For Use With (Equipment) pH Meter and Electrode
pH 4.01 ± 0.01 pH
Physical Form Liquid
Quantity 5 x 60 mL
Solution Type pH Buffer
Formulation Water, Potassium Hydroxide, Amaranth Red No. 27, Potassium Hydrogen Phthalate
Traceability to NIST Yes
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.