Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ pH Buffer Cubitainers™

Catalog No. 1330169
Change view
Click to view available options
Quantity:
19 L
pH:
4.01
7
10.01
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity pH
13-301-69 19 L 4.01
13-301-70 19 L 7
13-301-71 19 L 10.01
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 13-301-69 Supplier Thermo Scientific™ Supplier No. 9104CB
Add to Cart
Edge
Add to Cart
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Hold 5 gallons (19L) of pH buffer in color-coded Thermo Scientific™ Orion™ pH Buffer Cubitainers™ with easy-pour spigot.

Hold 5 gallons (19L) of pH buffer in color-coded Thermo Scientific™ Orion™ pH Buffer Cubitainers™ with easy-pour spigot.

  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST™ traceability
  • Manufactured under ISO 9000 quality standards
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material pH Buffer, 5 gallon cubitainers
Boiling Point 100°C
Color Light Red
Description pH 4.01 Buffer, 5 gallon
For Use With (Application) Accurate, reproducible pH calibrations
For Use With (Equipment) pH Meter and Electrode
pH 4.01
Physical Form Liquid
Quantity 19 L
Formulation Water, Potassium Hydroxide, Amaranth Red No. 27, Potassium Hydrogen Phthalate
Packaging Cubitainer
Traceability to NIST Yes
Solubility Information Soluble
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.