Learn More
Thermo Scientific™ Orion™ Potassium Ion Selective Electrodes
Catalog No. 133018
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
None
For Use With (Equipment):
Combination Electrode
Half-Cell Electrode
ISE Meter
Type:
Combination Electrode
Replacement Module
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types For Use With (Equipment) Type
13-301-8 None Combination Electrode Replacement Module
13-641-912 None Half-Cell Electrode Replacement Module
13-642-271 BNC (Waterproof) ISE Meter Combination Electrode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-301-8 Supplier Thermo Scientific™ Supplier No. 971901
Add to Cart
Add to Cart
Measure potassium ions in aqueous solutions simply, accurately and economically.
Measure potassium ions in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Potassium Electrodes.
  • Calibration Standard Catalog number: 0.1M KCl / 921906
  • Required ISA Catalog number: 931911
TRUSTED_SUSTAINABILITY
Specifications
Connector Types None
Description Replacement Module for Potassium Sure-Flow Combination Electrode 9719BNWP
Drift drift-free potentials
Junction Sure-Flow Junction
For Use With (Equipment) Combination Electrode
Type Replacement Module
Parameter Potassium