Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Powder Chemistry Packs

Catalog No. 13642214
Change view
Click to view available options
Type:
Chlorine (free)
Chlorine (total)
Iron (ferro)
Manganese
Molybdate/Molybdenum
Nitrite
Phosphate (ortho)
Silica
Sulfate
Range:
0.01 to 0.3 mg/L N
0.02 to 2 mg/L Cl2
0.02 to 3 mg/L Fe
0.06 to 2.5 mg/L PO4
0.1 to 18 mg/L Mn
0.5 to 66 mg/L MoO4
1 to 90 mg/L SiO2
5 to 100 mg/L SO4
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Range
13-642-214 Phosphate (ortho) 0.06 to 2.5 mg/L PO4
13-642-215 Nitrite 0.01 to 0.3 mg/L N
13-642-217 Manganese 0.1 to 18 mg/L Mn
13-642-213 Silica 1 to 90 mg/L SiO2
13-642-220 Chlorine (free) 0.02 to 2 mg/L Cl2
13-642-211 Chlorine (total) 0.02 to 2 mg/L Cl2
13-642-219 Iron (ferro) 0.02 to 3 mg/L Fe
13-642-212 Sulfate 5 to 100 mg/L SO4
13-642-216 Molybdate/Molybdenum 0.5 to 66 mg/L MoO4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13-642-214 Supplier Thermo Scientific™ Supplier No. AC4P95
Add to Cart
Add to Cart

Achieve easier and cleaner measurements with pre-measured and ready-to-use powder chemistry packs.

Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.

  • Equivalent to other widely used and recognized chemistries
  • Compatible with Orion™ AQUAfast™ Colorimeters
TRUSTED_SUSTAINABILITY

Specifications

Type Phosphate (ortho)
EPA Approved Methods Drinking Water: ASTM D515-88 (A) / Standard Methods 4500-P E; Wastewater: Standard Methods 4500-P E
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter
Includes 100 Phos3 Powder Packs
Packaging Powder Pack
Quantity 100 tests
Range 0.06 to 2.5 mg/L PO4
Recommended Accessories AQUAfast 24 mm round vials (Cat. No. AC2V24)
Test Method Phosphomolybdenum/Ascorbic Acid

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.