Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor

Catalog No. 13641164
Change view
Click to view available options
Chemical Name or Material:
Low Ionic Strength pH Test Kit
pH Buffer A
pH Buffer B
pH Buffer C
pHISA Low Ionic Strength Adjustor
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
13-641-164 pH Buffer A
13-641-165 pH Buffer B
13-641-722 pHISA Low Ionic Strength Adjustor
13-641-166 pH Buffer C
13-641-735 Low Ionic Strength pH Test Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13-641-164 Supplier Thermo Scientific™ Supplier No. 700702
Add to Cart
Add to Cart

Sample well, rain, distilled/deionized, boiler feed and process waters with this adjustor.

Sample well water, rain water, distilled/deionized water, boiler feed water, process water and more with the Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor. It offers fast, accurate pH results in high purity and low ionic strength waters.

Features:

  • Increases measurement accuracy
  • Reduces electrode drift
  • Eliminates contamination of sample

Pure Water pH Test Kit:

  • Kit contains special low ionic strength buffers for calibration and pHISA™ adjustor additive, to increase sample ionic strength without changing its pH
  • For best results, a high quality pH electrode such as the Thermo Scientific™ Orion™ ROSS Ultra™ glass-body pH electrode, Cat. No. 8102BNUWP, is recommended
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material pH Buffer A
Boiling Point 100°C
Color Clear
For Use With (Application) Provide fast, accurate pH results in high purity and low ionic strength waters. Recommended for samples such as well water, rain water, distilled/deionized water, boiler feed water and process water.
For Use With (Equipment) pH Meter and Electrode
pH 6.97
Physical Form Liquid
Quantity 4 x 475 mL
Packaging Bottle
Solubility Information Soluble

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.