Learn More
Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes PROMO
Description
Unique internal element system replaces Ag/AgCl or Hg2Cl2 internals with platinum wire and redox-couple filling solution. Double-junction reference cells have porous ceramic or glass fiber outer junction. Design provides stable potential, independent of temperature, and stable readings even when samples vary by 50°C (90°F) or more. Minimizes drift, stirring effects, clogged junctions; no need for frequent standardization so samples are analyzed faster.
- Epoxy body for durability and ruggedness
- Built-in ATC probe
- General-purpose applications
- Standard size and standard bulb
- Glass fiber junction
- L × Dia.: 120 × 12 mm
Specifications
- 0 to 14 pH
- 0 to 100°C (32 to 212°F) range
- Slope, 92 to 102%; isopotential point, pH 7
- When standardized in 6.86 pH buffer at 25°C (77°F), accuracy is ±0.03 pH for the buffer from 0 to 100°C (32 to 212°F) (using ATC); response is stable to 0.01 pH within 30 seconds at 25°C (77°F) and 75°C (167°F) and stable to ±0.002 pH within 15 seconds when cycling between pH 6.86 and 4.01 buffers at 25°C (77°F)
- Max. cap dia. (top): 0.75 in. (19 mm); collar dia. 0.62 in. (16 mm); Cable length is 39.37 in. (1 m)
Specifications
Specifications
| Description | Orion ROSS Ultra Refillable pH/ATC Triode Combination Electrodes with a BNC and 8-pin MiniDIN Connector and 1 meter cable |
| Connector Types | BNC (Waterproof), 8-pin Mini-DIN |
| Junction | Glass Fiber |
| Material | Epoxy Body |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 100°C |
| Type | pH/ATC Combination Electrode |
| Filling Solution Type | ROSS electrode 3 M KCl fill solution (Cat. No. 810007) |
| For Use With (Equipment) | Orion Versa Star, Star A and Dual Star Meters |
| Internal Reference | ROSS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.