Learn More
Thermo Scientific™ Orion Star™ A111 Benchtop pH Meter
Description
Take simple, routine pH, mV and temperature measurements with the Thermo Scientific™ Orion Star™ A111 Benchtop pH Meter. This meter is an easy-to-use and budget-friendly instrument for basic pH analysis in the lab. Perform up to a 3 point pH calibration with easy recall of calibration points and slope. Quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.
Features:
- Large, informative screen clearly shows the main information, including pH or mV reading, temperature, meter mode, calibration information and power source.
- Thermo Scientific™ AUTO-READ™ locks in the stable reading on your screen and ready indicator alerts when readings are stable.
- Meter offers up to 3 point pH calibration and automatic recognition of USA/NIST and DIN buffers.
- Use can easily recall calibration slope data for procedure checks.
- Nonvolatile memory holds up to 50 data points.
- Included electrode arm and newly designed probe holder make it easier to maintain and place probes into samples.
- Plug the meter into almost any power outlet with the included universal power adapter or use four AA batteries (sold separately) to run the meter on DC power.
- The IP54-rated housing can handle splashes and can be wall-mounted if table space is a problem.
Includes:
Orion STAR: A111 Benchtop pH Meter, Part no. STARA1110, includes electrode arm with redesigned holder and universal power adapter
Orion STAR: A111 Benchtop pH Meter Kit, Part no. STARA1115, includes 9157BNUMD Orion Triode™ 3-in-1 pH/ATC probe; Solution kit (60mL each of pH 4, 7 and 10 buffers; storage solution and cleaning solution); electrode arm with redesigned holder and universal power adapter
Orion STAR: A111 Benchtop pH Meter Kit, Part no. STARA1116, includes GD9156BNWP Orion Green electrode; 927007MD Stainless Steel ATC Probe; electrode arm with redesigned holder and universal power adapter
Specifications
Specifications
| Description | Orion Star A111 benchtop pH meter with stand |
| Accuracy (Temperature) | ±0.1°C |
| Data Management Details | Internal Data Point Memory |
| Accuracy (mV) | ±0.2 or ±0.05%, whichever is greater |
| Accuracy (pH) | ±0.01 |
| Outputs | None |
| Includes | Star A111 pH meter, electrode stand, 100-240V universal power adapter |
| Measurement Modes | pH, mV, Relative mV with Temperature |
| Calibration | pH, relative mV (RmV) and temperature |
| Range (pH) | -2 to 16 |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.