Learn More
Platypus Technologies LLC Platypus Technologies Cell Migration Assay (FLEX), 96-Well Plate (x4) + 96 Stoppers (Not Inserted), Tissue-Culture Treated, 1 Kit
SDP
Supplier:  Platypus Technologies LLC CMAUFL4
Encompass_Preferred
Oris™ Cell Migration Assay kits use a 96-well plate with “stopper” barriers that create a central cell-free detection zone for cell migration experiments. Removing the stoppers allows the cells to migrate into the detection zone at the center of each well. The Oris™ Cell Migration Assay provides consistent detection zones to give you superior reproducibility, accuracy and precision in your experiments. Ideal for publication-ready, high-quality data with fewer samples. Use the Oris™ Cell Migration Assay to identify cell migration inhibitors and promoters for drug discovery, wound healing, or cancer research. FLEX Kit includes (4) 96-well plates (black, clear bottom), (96) Stoppers, (1) detection mask for plate reader analysis and (1) stopper removal tool. Plate is tissue-culture treated.
Catalog No. 50-135-5507
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.