Learn More
Platypus Technologies LLC Platypus Technologies Detection Mask, Accessory for Cell Migration Assay
SDP
Supplier:  Platypus Technologies LLC CMA.MM
Encompass_Preferred
Oris™ Detection Mask aids analysis of the Oris™ Cell Migration Assay plates in association with fluorescent plate readers to enhance data acquisition and reduce CVs and signal-to-noise ratio. Oris™ Detection Mask, CMA.MM
Catalog No. 50-135-5526
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.