Learn More
Platypus Technologies LLC Platypus Technologies Cell Migration Assay (UNIVERSAL), (5) 96-Well Plates + (480) Stoppers (Not Inserted), Tissue-Culture Treated
Supplier: Platypus Technologies LLC CMAU505
Oris™ Cell Migration Assay kits use a 96-well plate with “stopper” barriers that create a central cell-free detection zone for cell migration experiments. Removing the stoppers allows the cells to migrate into the detection zone at the center of each well. The Oris™ Cell Migration Assay provides consistent detection zones to give you superior reproducibility, accuracy and precision in your experiments. Ideal for publication-ready, high-quality data with fewer samples. Use the Oris™ Cell Migration Assay to identify cell migration inhibitors and promoters for drug discovery, wound healing, or cancer research. UNIVERSAL Kit includes (5) 96-well plates (black, clear bottom), (480) Stoppers, (1) detection mask for plate reader analysis and (1) stopper removal tool. Plates are tissue-culture treated.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.