Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orlistat, 98%

Catalog No. AAJ62999MC
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62999MC 100 mg
AAJ62999MF 50 mg
AAJ62999MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ62999MC Supplier Thermo Scientific Chemicals Supplier No. J62999MC
Add to Cart

CAS: 96829-58-2 | C29H53NO5 | 495.75 g/mol

Orlistat, used in obesity research, is a pancreatic lipase inhibitor that acts locally in the gastrointestinal tract to inhibit lipase. More detailed experiments show that gradually THL affects the retinoblastoma protein pathway and leads to cell-cycle arrest at G1/S boundary. Lipase Inhibitor, THL is an inhibitor of pancreatic and gastric lipase, as well as carboxyl ester lipase (CEL) and diacylglycerol lipase-α (DAGLα). Lipase Inhibitor, THL also activates caspase-3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Orlistat, used in obesity research, is a pancreatic lipase inhibitor that acts locally in the gastrointestinal tract to inhibit lipase. More detailed experiments show that gradually THL affects the retinoblastoma protein pathway and leads to cell-cycle arrest at G1/S boundary. Lipase Inhibitor, THL is an inhibitor of pancreatic and gastric lipase, as well as carboxyl ester lipase (CEL) and diacylglycerol lipase-α (DAGLα). Lipase Inhibitor, THL also activates caspase-3.

Solubility
Soluble in DMSO (19 mg/mL), ethanol (20 mg/mL), and DMF (20 mg/mL).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96829-58-2
Molecular Formula C29H53NO5
Molecular Weight (g/mol) 495.75
MDL Number MFCD05662360
InChI Key AHLBNYSZXLDEJQ-FWEHEUNISA-N
Synonym orlistat, tetrahydrolipstatin, xenical, alli, orlipastat, --tetrahydrolipstatin, orlipastatum, orlipastatum inn-latin, thlp, tetrahydrolipastatin
PubChem CID 3034010
IUPAC Name (2S)-1-[(2S,3S)-3-hexyl-4-oxooxetan-2-yl]tridecan-2-yl (2S)-2-formamido-4-methylpentanoate
SMILES CCCCCCCCCCC[C@@H](C[C@@H]1OC(=O)[C@H]1CCCCCC)OC(=O)[C@H](CC(C)C)NC=O

Specifications

Melting Point <50°C
Color White
Quantity 100 mg
Merck Index 14,6869
Solubility Information Soluble in DMSO (19mg/mL),ethanol (20mg/mL),and DMF (20mg/mL).
Formula Weight 495.73
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Orlistat
RTECSNumber OH3167600
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.