Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orlistat 97.0+%, TCI America™
SDP

Catalog No. O0381100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0381100MG 100 mg
O03811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0381100MG Supplier TCI America Supplier No. O0381100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 96829-58-2
Molecular Formula C29H53NO5
Molecular Weight (g/mol) 495.75
MDL Number MFCD05662360
InChI Key AHLBNYSZXLDEJQ-FWEHEUNISA-N
Synonym orlistat, tetrahydrolipstatin, xenical, alli, orlipastat, --tetrahydrolipstatin, orlipastatum, orlipastatum inn-latin, thlp, tetrahydrolipastatin
PubChem CID 3034010
IUPAC Name (2S)-1-[(2S,3S)-3-hexyl-4-oxooxetan-2-yl]tridecan-2-yl (2S)-2-formamido-4-methylpentanoate
SMILES CCCCCCCCCCC[C@@H](C[C@@H]1OC(=O)[C@H]1CCCCCC)OC(=O)[C@H](CC(C)C)NC=O

Specifications

Melting Point 45°C
Color White
Quantity 100 mg
Formula Weight 495.75
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Orlistat
RTECSNumber OH3167600
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.