Learn More
CPC Scientific H-Glu-Thr-Pro-Asp-Cys-Phe-Trp-Orn-Tyr-Cys-Val-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific UROT004A
Encompass
A cyclic peptide that has been discovered to be the endogenous ligand of an orphan GPCR and subsequently has been named the UT receptor. No synthetic ligands are available for investigating this novel peptide-receptor system. ONE-LETTER SEQUENCE: ETPDCFW-Orn-YCV (Cys5 and 10 bridge)MOLECULAR FORMULA: C63H83N13O18S2MOLECULAR WEIGHT:1374.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [479065-85-5], RESEARCH AREA: Cardiovascular
Catalog No. 50-211-0603