Learn More
ATCC Oropouche virus control ascitic fluid - positive control for assay validation (ATCC VR-1228CAF)
Supplier:  ATCC VR1228CAF
Encompass
Oropouche virus control ascitic fluid is a freeze-dried, mouse-derived ascitic preparation intended for use as a positive control in laboratory virology assays for Oropouche virus. This material is supplied for research use only and is not for human or animal therapeutic use, consumption, or diagnostic purposes. Handle and store according to the product information sheet and Certificate of Analysis.
  • Freeze-dried mouse ascitic fluid for stable storage.
  • Intended as a positive control in Oropouche virus assays.
  • Supplied for laboratory research use only.
  • Biosafety level 1 recommended for handling.
  • Viability warranted for 30 days from shipment when stored per instructions.
Catalog No. NC3136056