Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orotic Acid Monohydrate 98.0+%, TCI America™
SDP

Catalog No. O0065100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0065100G 100 g
O006525G 25 g
O0065500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O0065100G Supplier TCI America Supplier No. O0065100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50887-69-9
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.10
MDL Number MFCD00006027
InChI Key PXQPEWDEAKTCGB-UHFFFAOYSA-N
Synonym orotic acid monohydrate, orotic acid hydrate, 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, lactinium, oroticum acidum, 2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid, orotc acd hydrate?, uracil-6-carboxylic acid monohydrate, uracil-4-carboxylic acid hydrate
PubChem CID 16218504
IUPAC Name 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid
SMILES OC(=O)C1=CC(=O)NC(=O)N1

Specifications

Color White
Quantity 100 g
Formula Weight 156.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Orotic Acid Monohydrate
EINECSNumber (9)-0278
RTECSNumber RM3180000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.