Learn More
QuidelOrtho ABO Blood Group Reagents
Description
- Anti-A and Anti-B BioClone™ reagents are used to determine ABO cell type
- Provide the strong reactivity and product consistency associated with monoclonal technology
- Reagents are Murine Monoclonal blends
- Anti-A,B BioClone reagent is a blend of Anti-A and Anti-B monoclonal antibodies used for ABO group confirmation; can be used by transfusion services to confirm blood group O donor units
- Anti-A and Anti-A,B can detect many A subgroups (such as A3, Ax, or AsubB)
Specifications
Specifications
| Type | Anti-A, B BioClone™ |
| For Use With (Application) | To determine ABO cell type |
| Quantity | 15 x 10 mL |