Learn More
QuidelOrtho BioClone™ Reagents
Description
Anti Sera products provide qualitative tests for recognition of antigens on human red blood cells
- Using high-protein reagents prepared from human serum, Anti-D blood grouping reagent recognizes D (Rh0) antigens
- Anti-C (Anti-RH2), Anti-E (Anti-RH3), Anti-c (Anti-RH4) and Anti-e (Anti-RH5) blood grouping reagents are low-protein reagents prepared from human/mouse cell lines; used to recognize corresponding antigens on human red blood cells
- The control is prepared from pools of human serum tested and found to be negative for blood group antibodies
Specifications
Specifications
| Type | Anti-e BioClone™ Reagent |
| For Use With (Application) | To recognize spontaneous Agglutination of red Blood cells |
| Quantity | 1 x 5 mL |