Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho MicroTip™ Supplies Diluent Packs for VITROS™ 5,1 FS System

Catalog No. OCD6802326
Change view
Click to view available options
Quantity:
6 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OCD6802326 6 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OCD6802326 Supplier QuidelOrtho Supplier No. 6802326
Add to Cart
Add to Cart

Specifications

Chemical Name or Material FS diluent pack 4 (DAT diluent 1/ DAT diluent 2)
For Use With (Application) Used for dilution of calibrators and samples for drug abuse assays
For Use With (Equipment) Vitros™ 5, 1FS chemistry system
Quantity 6 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.