Learn More
Description
Precision and accuracy
- Uses proven reliable ID-MTS™ Gel Cards
- Validated system of ID-MTS™ Gel Card with centrifuge and incubator
- Clear, stable, well-defined end points
- Objective interpretation of results and full assay portfolio
Convenient, compact 2 in 1 QuidelOrtho Workstation
- Integrated system of ID-MTS™ Gel Cards, centrifuge and incubator
- Light weight and small footprint for space-constrained labs
- Labs can standardize on one technology
Improved Process Controls
- Incubator temperature monitoring with visual indicator
- Unique beeping pattern to indicate cycle completion
- Enhanced error code intelligence for troubleshooting
- Quiet centrifuge with translucent panels for visual monitoring
Ease of Use
- Language-neutral icons and enhanced alarm systems
- Easy to perform centrifuge QC using a tachometer
- Quick access to holding area for centrifuge balance-card
Designed with the environment in mind
- Reduction in device weight by 33%
- Reduction in external packaging by 54%
- Reduction in Reference Guide size by 22%
- Reduced energy consumption by 70%
Applications:
- ABO Grouping/Rh Testing
- NeoNatal Blood Type (Forward Only)
- Blood Unit Confirmation (Group A, B, AB)
- Blood Unit Confirmation (Group O)
- Indirect Antiglobulin Testing
- Compatibility Testing (Coombs Crossmatch and Immediate Spin Crossmatch)
- Direct Antiglobulin Testing
- Rh Phenotyping
- Rh Typing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Includes | Workstation with ID-TipMaster™ Pipettor |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

