Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ High Sample Diluents: B-Manual

Catalog No. OCD1435866
Change view
Click to view available options
Quantity:
4 x 27 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OCD1435866 4 x 27 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OCD1435866 Supplier QuidelOrtho Supplier No. 1435866
Add to Cart
Add to Cart

Specifications

Chemical Name or Material High sample diluent B (Buffered)
For Use With (Equipment) Vitros™ Myoglobin assay
Quantity 4 x 27 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.