Learn More
QuidelOrtho VITROS™ MicroSlides
Specifications
Specifications
| Sample Type | Heparin, Plasma, Serum, Urine |
| Type | Magnesium Dry Slide |
| Formulation | 1,2-bis(o-Aminophenoxy)ethane-N,N,N',N'-Tetraacetic Acid (Calcium chelator) 242μg, 1,5-bis(2-Hydroxy-3,5-Dichlorophenyl)-3-Cyanoformazan (dye) 38μg, Pigment, Binders, Buffer, dye solubilizer, Surfactants, cross-linking |
| For Use With (Application) | In-vitro diagnostic use |
| Quantity | 5 x 60 Pack |
| Sample Size | 5 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.