Learn More
QuidelOrtho VITROS™ MicroSlides
Specifications
Specifications
| Sample Type | Heparin, Plasma, Serum |
| Type | Total Protein Dry Slide |
| Formulation | Copper Sulfate 0.9mg, Tartaric Acid 1.2mg and Lithium Hydroxide 1.3mg, Polymer beads, Binders and Surfactants |
| For Use With (Application) | In-vitro diagnostic use |
| Quantity | 5 x 50 Pack |
| Sample Size | 10 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.