Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ortho-Clinical Diagnostics™ VITROS™ MicroSlides
SDP

Catalog No. 23046174 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
VITROS 250 Dry Slides: Magnesium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-046174 VITROS 250 Dry Slides: Magnesium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046174 Supplier Ortho-Clinical Diagnostics™ Supplier No. 8255093

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with the Ortho VITROS™ 250 System

Specifications

Sample Type Heparin, Plasma, Serum, Urine
Type Magnesium Dry Slide
Formulation 1,2-bis(o-Aminophenoxy)ethane-N,N,N',N'-Tetraacetic Acid (Calcium chelator) 242μg, 1,5-bis(2-Hydroxy-3,5-Dichlorophenyl)-3-Cyanoformazan (dye) 38μg, Pigment, Binders, Buffer, dye solubilizer, Surfactants, cross-linking
For Use With (Application) In-vitro diagnostic use
Quantity 5 x 60 Pack
Sample Size 5 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.