Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ MicroTip™ Diluent Packs

Catalog No. OCD6801753
Change view
Click to view available options
Chemical Name or Material:
FS Diluent Pack 1 (Apo Diluent/UED)
FS Diluent Pack 2 (BSA/Saline)
FS Diluent Pack 3 (Specialty Diluent/water)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
OCD6801753 FS Diluent Pack 2 (BSA/Saline)
OCD6801752 FS Diluent Pack 1 (Apo Diluent/UED)
OCD6801754 FS Diluent Pack 3 (Specialty Diluent/water)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. OCD6801753 Supplier QuidelOrtho Supplier No. 6801753
Add to Cart
Add to Cart

Specifications

Chemical Name or Material FS Diluent Pack 2 (BSA/Saline)
For Use With (Equipment) Vitros™ 5, 1FS chemistry system
Quantity 3 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.