Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ MicroTip™ Reconstitution Diluent

Catalog No. OCD6801874
Change view
Click to view available options
Quantity:
12 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OCD6801874 12 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OCD6801874 Supplier QuidelOrtho Supplier No. 6801874
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material Reconstitution diluent
For Use With (Equipment) Vitros™ 5, 1FS chemistry system
Quantity 12 Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.