Learn More
QuidelOrtho Optix™ Reader
SDP
Catalog No. OK6842222 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
For Use With (Application):
Automating Reaction Grading
Catalog No. For Use With (Application)
OK6842222 Automating Reaction Grading
1 options
This item is not returnable. View return policy
Catalog No. OK6842222 Supplier QuidelOrtho Supplier No. 6842222
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to automate reaction grading results interpretation and data management, provides complete semi-automated solution when paired with QuidelOrtho Workstation and QuidelOrtho ID-MTS gel technology
  • For direct and indirect antiglobulin testing, as determined by external validation testing using BioVue™ technology
    • Acceptance criteria for concordance for direct and indirect antiglobulin: 98.0%, lower bound 95% CI of 99.7%
  • Ensures Accurate Results:
    • Shown to deliver 99.9% concordance, same high quality and consistent results as automated solution QuidelOrtho VISION™ Analyzer
  • Eliminates Guesswork:
    • Uses high resolution images of each side of reaction column to determine result
      • Eliminates subjectivity and discrepancies associated with human reading of results
  • Minimizes Manual Error:
    • Delivers results directly to computer or from bi-directional interface of lab's LIS
      • Reduces or eliminates need for manual documentation
      • Eliminates risk of transcription errors
Specifications
Type Advanced Card Reader
For Use With (Application) Automating Reaction Grading