Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ortho-Phosphoric acid 85%, Suprapur™, MilliporeSigma™

Catalog No. M1005521000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1005521000 1 L
M1005520250 250 mL
M1005520500 500 mL
M1005522500 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1005521000 Supplier MilliporeSigma Supplier No. 1.00552.1000
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid

Specifications

Color Colorless
Melting Point 21°C
Boiling Point 158°C
Density 1.71g/cm3 (20°C)
Packaging Plastic Bottle
Vapor Pressure 2 hPa (20°C)
Assay Percent Range 85%
Quantity 1 L
Physical Form Liquid
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
Solubility Information Soluble (2°C)
SMILES OP(=O)(O)O
Molecular Weight (g/mol) 97.994
Grade Suprapur
Chemical Name or Material Suprapur™
Show More Show Less
Hazard Category SignalWord: Danger
Recommended Storage Store above +15°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.