Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho Reagent Red Cells
SDP

Catalog No. 23287467
Change view
Click to view available options
Quantity:
1 x 10 mL
10 mL
2 x 10 mL
3 x 10 mL
4 x 5 mL
Each
Type:
Affirmagen™
Affirmagen™ 3 Reagent Red Cells
Affirmagen™ Reagent Red Cells
Alba Q-Chek™ Simulated Whole Blood Control
Coombs Control Reagent Red Cells
Fetalscreen™ II
Pooled Screening Reagent Red Cells
SelectogeN™ Reagent Red Cells
Surgiscreen™ Reagent Red Cells
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-287-467 10 mL Pooled Screening Reagent Red Cells
23-287-331 10 mL Surgiscreen™ Reagent Red Cells
23-287-316 10 mL SelectogeN™ Reagent Red Cells
23-287-330 1 x 10 mL Coombs Control Reagent Red Cells
23-287-325 2 x 10 mL Affirmagen™ Reagent Red Cells
23-287-501 2 x 10 mL Affirmagen™
23-287-463 3 x 10 mL Affirmagen™ 3 Reagent Red Cells
23-287-469 Each Alba Q-Chek™ Simulated Whole Blood Control
23-287-468 4 x 5 mL Fetalscreen™ II
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
All blood products should be treated as potentially infectious. Source material from which this product was derived was found negative when tested in accordance with current FDA required tests. No known test methods can offer assurance that products derived from human blood will not transmit infectious agents.
Catalog No. 23-287-467 Supplier QuidelOrtho Supplier No. 6902314
Add to Cart
Add to Cart
All blood products should be treated as potentially infectious. Source material from which this product was derived was found negative when tested in accordance with current FDA required tests. No known test methods can offer assurance that products derived from human blood will not transmit infectious agents.

Reagent red blood cells are used in the detection and identification of a variety of blood group antibodies

  • Tests include hemagglutination and hemolysis-based results
  • A variety of testing functions are available, including:
    • Detection and/or identification of unexpected blood group antibodies
    • Affirmation of expected group antibodies
    • Serum grouping
    • Confirmation of negative antiglobulin tests
  • Screening for RhD (Rh0) positive fetal red blood cells

Specifications

Type Pooled Screening Reagent Red Cells
For Use With (Application) For detection and identification of a variety of Blood group Antibodies
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.