Learn More
CPC Scientific H-Tyr-Leu-Gly-Ala-Ser-Val-Pro-Ser-Pro-Asp-Pro-Leu-Glu-Pro-Thr-Arg-Glu-Gln-Cys-Glu-Leu-Asn-Pro-Ala-Cys-Asp-Glu-Leu-Ser-Asp-Gln-Tyr-Gly-Leu-Lys-Thr-Ala-Tyr-Lys-Arg-Ile-Tyr-Gly-Ile-Thr-Ile-OH
(Disulfide bond) 0.5MG
SDP
Supplier:  CPC Scientific OSTP010A
Encompass
This peptide is a bone-specific hormone involved in the regulation of energy metabolism, for which the Gla peptide acts as a precursor and transport form.SEQUENCE: H-Tyr-Leu-Gly-Ala-Ser-Val-Pro-Ser-Pro-Asp-Pro-Leu-Glu-Pro-Thr-Arg-Glu-Gln-Cys-Glu-Leu-Asn-Pro-Ala-Cys-Asp-Glu-Leu-Ser-Asp-Gln-Tyr-Gly-Leu-Lys-Thr-Ala-Tyr-Lys-Arg-Ile-Tyr-Gly-Ile-Thr-Ile-OH(Disulfide bond)ONE-LETTER SEQUENCE: YLGASVPSPDPLEPTREQCELNPACDELSDQYGLKTAYKRIYGITI (Cys & Cys bridge)MOLECULAR FORMULA: C226H351N57O74S2MOLECULAR WEIGHT: 5114.75STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [1802086-27-6]SYNONYMS: (Glu)-Osteocalcin (1-46) (mouse)RESEARCH AREA: OsteoporosisREFERENCES:N.K.Lee et al., Cell, 130, 456 (2007)
SKU(s): OSTP-010A, OSTP-010B
Catalog No. 50-211-0015
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.