Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ OSBP1 Peptide
SDP

Catalog No. NB803598 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB803598 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB803598 Supplier Novus Biologicals™ Supplier No. NB1001500PEP
Add to Cart
Add to Cart

Specifications

Gene ID (Entrez) 5007
Species Human
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-1500
Gene Alias OSBP1oxysterol-binding protein 1, oxysterol binding protein
Gene Symbol OSBP
Quantity 0.1 mg
Research Category Lipid and Metabolism
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.